Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 3/CD61 Antibody (F11), FITC, Novus Biologicals™
SDP

Catalog No. NB10063958 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063958 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10063958 Supplier Novus Biologicals Supplier No. NB10063958

Mouse Monoclonal Antibody

Integrin beta 3/CD61 Monoclonal specifically detects Integrin beta 3/CD61 in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Integrin beta 3/CD61
Applications Flow Cytometry
Classification Monoclonal
Clone F11
Concentration 0.1 mg/mL
Conjugate FITC
Dilution Flow Cytometry 1:10-1:1000
Gene Alias CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa
Gene Symbols ITGB3
Host Species Mouse
Immunogen Bone cell suspension from SD rats
Purification Method Protein A or G purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cancer, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3690
Test Specificity NB100-65387 recognises the rat CD61 cell surface antigen, also known as the integrin b3 chain. CD61 is expressed by platelets, megakaryocytes and osteoclasts.
Target Species Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.