Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 3/CD61 Antibody (ITGB3/2166R), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB392524 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392524 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392524 Supplier Novus Biologicals Supplier No. NBP308291AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Integrin beta 3/CD61 Monoclonal antibody specifically detects Integrin beta 3/CD61 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Integrin beta 3/CD61
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ITGB3/2166R
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa
Host Species Rabbit
Immunogen Recombinant human Integrin beta 3/CD61 protein fragment (aa 385-490) (exact sequence is proprietary) (Uniprot: P05106)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3690
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.