Learn More
Description
Specifications
Specifications
| Antigen | Integrin beta 3/CD61 |
| Applications | Flow Cytometry, Neutralization, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | PM6/13 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry <= 1 ug/10^6 cells, Block/Neutralize |
| Gene Accession No. | P05106 |
| Gene Alias | CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa |
| Gene Symbols | ITGB3 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
