Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 3/CD61 Antibody (VIPL2), FITC, Novus Biologicals™
SDP

Catalog No. NB355791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355791 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355791 Supplier Novus Biologicals Supplier No. NBP197563F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin beta 3/CD61 Monoclonal antibody specifically detects Integrin beta 3/CD61 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF

Specifications

Antigen Integrin beta 3/CD61
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF
Classification Monoclonal
Clone VIPL2
Conjugate FITC
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, CyTOF-ready
Formulation PBS
Gene Alias CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa
Host Species Mouse
Immunogen The mouse monoclonal antibody VIPL2 recognizes CD61
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3690
Target Species Human, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.