Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 3/CD61, p Tyr785 Antibody, Novus Biologicals™
SDP

Catalog No. NB017851 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017851 0.025 mL
NB10081995 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017851 Supplier Novus Biologicals Supplier No. NB100819950.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Integrin beta 3/CD61 (p Tyr785) Polyclonal specifically detects Integrin beta 3/CD61 (p Tyr785) in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Integrin beta 3/CD61 (p Tyr785)
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
Gene Accession No. P05106
Gene Alias CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa
Gene Symbols ITGB3
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from human Integrin 3 around the phosphorylation site of tyrosine 785 (I-T-YP-R-G).
Molecular Weight of Antigen 110 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cytoskeleton Markers, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3690
Test Specificity Integrin 3 (specific to Phospho-Tyr785) detects endogenous levels of Integrin 3 only when phosphorylated at tyrosine 785.
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.