Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 4/CD104 Antibody (439-9B), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP143369C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143369C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143369C Supplier Novus Biologicals Supplier No. NBP143369C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin beta 4/CD104 Monoclonal specifically detects Integrin beta 4/CD104 in Human, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Integrin beta 4/CD104
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 439-9B
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4
Gene Symbols ITGB4
Host Species Rat
Immunogen The immunogen for this antibody was CD104.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3691.0
Target Species Human, Bovine
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.