Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 4/CD104 Antibody (450-9D) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10065599 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065599 0.1 mg
NB165599SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10065599 Supplier Novus Biologicals Supplier No. NB10065599
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Integrin beta 4/CD104 Monoclonal specifically detects Integrin beta 4/CD104 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.

Specifications

Antigen Integrin beta 4/CD104
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 450-9D
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:25-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:500-1:1000
Gene Accession No. P16144
Gene Alias CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4
Gene Symbols ITGB4
Host Species Mouse
Immunogen Purified alpha6 beta4 integrin from A431 cells
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3691
Test Specificity NB100-65599 recognizes the human beta4 integrin (also known as CD104), a 205kD glycoprotein which associates with the alpha6 integrin to form the alpha6/beta4 complex. CD104 is expressed on epithelial cells, Schwann cells and various tumour cell lines. NB100-65599 recognizes an extracellular epitope on the CD104 molecule.
Target Species Human, Ferret
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.