Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 4/CD104, p Tyr1494 Antibody, Novus Biologicals™
SDP

Catalog No. NB387850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387850 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387850 Supplier Novus Biologicals Supplier No. NBP323397
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Integrin beta 4/CD104 Polyclonal antibody specifically detects Integrin beta 4/CD104 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Integrin beta 4/CD104
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunocytochemistry/ Immunofluorescence 1:100
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4
Host Species Rabbit
Immunogen Phospho-Integrin beta 4/CD104 (Tyr-1494) synthetic peptide (coupled to KLH) corresponding to amino acid residues around tyrosine 1494 of human Integrin beta 4/CD104. This peptide sequence is found in all three Integrin beta 4/CD104 isoforms and is highly conserved in rat and mouse Integrin beta 4/CD104. [UniProt# P16144-2]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3691
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.