Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 6 Antibody (268622), PerCP, Novus Biologicals™
SDP

Catalog No. NB304481 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304481 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304481 Supplier Novus Biologicals Supplier No. FAB2389C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin beta 6 Monoclonal specifically detects Integrin beta 6 in Mouse samples. It is validated for Immunocytochemistry, Western Blot.

Specifications

Antigen Integrin beta 6
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 268622
Conjugate PerCP
Dilution Western Blot, Immunocytochemistry
Formulation PBS
Gene Alias integrin beta-6, integrin, beta 6
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 6, Gly22-Asn706, Accession # Q9Z0T9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3694
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.