Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 6 Antibody (437211), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB370792 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370792 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370792 Supplier Novus Biologicals Supplier No. FAB4155L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin beta 6 Monoclonal specifically detects Integrin beta 6 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Integrin beta 6
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 437211
Conjugate DyLight 550
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias integrin beta-6, integrin, beta 6
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha V beta 6, Phe31-Val992 (Integrin alpha V) and Gly22-Asn707 (Integrin beta 6), Accession # P18564
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3694
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.