Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 6 Antibody (OTI1D2), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP271051UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271051UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271051UV Supplier Novus Biologicals Supplier No. NBP271051UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin beta 6 Monoclonal specifically detects Integrin beta 6 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Integrin beta 6
Applications Western Blot
Classification Monoclonal
Clone OTI1D2
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias integrin beta-6, integrin, beta 6
Gene Symbols ITGB6
Host Species Mouse
Immunogen Full length recombinant protein of human Integrin beta 6 (NP_000879) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3694.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.