Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 7 Antibody (473207), HRP, Novus Biologicals™
SDP

Catalog No. NB347669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347669 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347669 Supplier Novus Biologicals Supplier No. FAB4669H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Integrin beta 7
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 473207
Conjugate HRP
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Integrin beta 7 long isoform, Glu20-His723, Accession # P26010
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3695
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.