Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 7, Rabbit anti-Human,Mouse, Clone: FIB27, Novus Biologicals™
SDP

Catalog No. NB015742 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015742 0.025 mg
NB015743 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB015742 Supplier Novus Biologicals Supplier No. NBP2810870.025MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Block/Neutralize.

Specifications

Antigen Integrin beta 7
Applications Flow Cytometry, Immunoprecipitation, Blocking Assay
Classification Monoclonal
Clone FIB27
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Immunoprecipitation, Block/Neutralize
Formulation PBS
Gene Alias Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7
Gene Symbols ITGB7
Host Species Rabbit
Immunogen FIB27 was prepared by immunizing rats with TK1 cells.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3695
Test Specificity FIB27 binds specifically to both human and mouse Integrin beta 7 at epitope region Dl on which is involved in all a4b7-mediated adhesion events. Does not compete with DAKT32 for binding. FIB27 blocks LS722-induced TK1 cell aggregation (mAb LS722 is an activating anti-a4B7 antibody and induces aggregation via an a4B7-dependent pathway). Integrin beta 7 is one subunit is a heterodimer which makes up an intergin molecule. In the mouse beta-7 is selectively found on the majority of mature lymphocytes, whereas a small subpopulation of thymocytes and bone marrow cells express beta-7. Beta-7 is also expressed in variable amounts in CD4 T memory cells. Integrin molecules mediate cell-cell and cell-extracellular matrix adhesion and are involved in lymphocyte homing, leukocyte recruitment to inflammatory sites, myogenesis, hemopoiesis, and melanoma metastasis.
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.