Learn More
Integrin beta 7, Rabbit anti-Human,Mouse, Clone: FIB27, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Integrin beta 7 |
| Applications | Flow Cytometry, Immunoprecipitation, Blocking Assay |
| Classification | Monoclonal |
| Clone | FIB27 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry, Immunoprecipitation, Block/Neutralize |
| Formulation | PBS |
| Gene Alias | Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7 |
| Gene Symbols | ITGB7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.