Learn More
Integrin beta 7 Antibody (2G18) - Azide and BSA Free, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NB11094147100UG
 View more versions of this product
Catalog No. NBT94147100
Only null left
Add to Cart
Description
Description
Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
Specifications
Integrin beta 7
Monoclonal
0.4 mg/mL
Flow Cytometry 1:50-200, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:100-400, Immunohistochemistry-Paraffin 1:100-200
Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7
Rat
Protein G purified
Signal Transduction
3695
Mouse
Purified
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
2G18
Unconjugated
A 0.2 um filtered solution in PBS with No Preservative
ITGB7
Cell lysates of mouse lymphocytes
100 μg
Primary
This was selected for its ability to detect mouse CD45 positive leukocytes in IHC.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only