Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 8 Antibody (2723C), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB296213 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296213 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296213 Supplier Novus Biologicals Supplier No. FAB47752X
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Integrin beta 8 Monoclonal specifically detects Integrin beta 8 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Integrin beta 8
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 2723C
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias integrin beta-8, integrin, beta 8
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line CHO-derived human Integrin alpha V beta 8, Human Integrin alpha V (Phe31-Val992) and Human Integrin beta 8 (Glu43-Arg684), Accession # NP_002201.1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3696
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.