Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta-like protein 1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP159169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159169 100 μL
NBP15916920 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP159169 Supplier Novus Biologicals Supplier No. NBP159169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Integrin beta-like protein 1 Polyclonal specifically detects Integrin beta-like protein 1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Integrin beta-like protein 1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. O95965
Gene Alias integrin, beta-like 1 (with EGF-like repeat domains), OSCPITGBL1, integrin beta-like 1, Osteoblast-specific cysteine-rich protein, Ten integrin EGF-like repeat domain-containing protein, ten integrin EGF-like repeat domains protein, TIEDintegrin beta-like protein 1
Gene Symbols ITGBL1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ITGBL1 (integrin, beta-like 1 (with EGF-like repeat domains)) The peptide sequence was selected from the N terminal of ITGBL1. Peptide sequence CSNAGTCHCGRCKCDNSDGSGLVYGKFCECDDRECIDDETEEICGGHGKC.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9358
Test Specificity Expected identity based on immunogen sequence: Human: 100%; Bovine: 92%; Equine: 92%; Pig: 92%; Rabbit: 92%; Chicken: 85%; Canine: 85%; Guinea pig: 85%; Mouse: 85%; Rat: 85%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.