Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta-like protein 1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP182473 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP182473 0.1 mL
NB407146 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP182473 Supplier Novus Biologicals Supplier No. NBP182473
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Integrin beta-like protein 1 Polyclonal specifically detects Integrin beta-like protein 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Integrin beta-like protein 1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias integrin, beta-like 1 (with EGF-like repeat domains), OSCPITGBL1, integrin beta-like 1, Osteoblast-specific cysteine-rich protein, Ten integrin EGF-like repeat domain-containing protein, ten integrin EGF-like repeat domains protein, TIEDintegrin beta-like protein 1
Gene Symbols ITGBL1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:VCGECTCHDVDPTGDWGDIHGDTCECDERDCRAVYDRYSDDFCSGHGQCNCGRCDCKAGWYGKKCEHPQSCTLSAEESIRKCQGSSDLPCSGRGKCECGKCTCYPPGDRRVYGKTCECDDRRCEDLDGV
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9358
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.