Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Intelectin-1/Omentin Mouse anti-Human, Clone: ITLN1/4065, Novus Biologicals™
SDP

Catalog No. NB122854 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB122854 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB122854 Supplier Novus Biologicals Supplier No. NBP30769020UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Intelectin-1/Omentin Monoclonal specifically detects Intelectin-1/Omentin in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen Intelectin-1/Omentin
Applications Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone ITLN1/4065
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Alias Endothelial lectin HL-1, FLJ20022, Galactofuranose-binding lectin, hIntL, HL-1, intelectin 1 (galactofuranose binding), intelectin-1, INTLHL1, ITLNITLN-1, LFRIntestinal lactoferrin receptor, omentin
Host Species Mouse
Immunogen Recombinant fragment (around aa16-141) of human Intelectin-1/Omentin protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55600
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.