Learn More
United Scientific Supplies Interferometer
SDPSCI_ED
Catalog No. S07392 Shop All United Scientific Supplies, Inc Products
Click to view available options
:
Interferometer
Determine the wavelength of light with great precision.
  • For Michelson or Fabry-Perot setups
  • High precision with interchangeable optics
  • 200mm precision-geared lead screw provides fine adjustment capability
Specifications
Product Type Interferometer
For Use With (Equipment) Michelson or Fabry-Perot setups
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.