Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Intermolecular Guanine Quadruplex DNA Antibody (1H6) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP24246825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24246825 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP24246825 Supplier Novus Biologicals Supplier No. NBP24246825UG

Mouse Monoclonal Antibody

Intermolecular Guanine Quadruplex DNA Monoclonal specifically detects Intermolecular Guanine Quadruplex DNA in Non-species specific samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Intermolecular Guanine Quadruplex DNA
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1H6
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS, pH7.4 with no preservative
Host Species Mouse
Immunogen Synthetic intermolecular guanine quadruplex DNA was used as the immunogen for this antibody.
Quantity 25 μg
Primary or Secondary Primary
Test Specificity synthetic intermolecular guanine quadruplex DNA
Reconstitution Reconstitute with 0.1 mL water.
Target Species Non-species specific
Content And Storage Lyophilized antibody can be kept at 4°C for up to 3 months and should be kept at -20°C for long-term storage. To avoid freeze-thaw cycles, reconstituted antibody should be aliquoted before freezing for short-term storage (-20°C) or for long-term storage (-80°C). For maximum recovery of product, centrifuge the original vial prior to removing the cap. Further dilutions can be made in assay buffer.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.