Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Intra Acrosomal Protein Antibody (Ds-2), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB500436G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500436G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500436G Supplier Novus Biologicals Supplier No. NB500436G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Intra Acrosomal Protein Monoclonal specifically detects Intra Acrosomal Protein in Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Intra Acrosomal Protein
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone Ds-2
Conjugate DyLight 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Gene Alias acrosomal protein SP-10, acrosomal vesicle protein 1D11S4365, SP-10, SPACA2, sperm protein 10
Gene Symbols ACRV1
Host Species Mouse
Immunogen Freshly ejaculated dog sperms were washed in PBS and extracted in 3% acetic acid, 10% glycerol, 30 mM benzaminidine. The acid extract was dialyzed against 0.2% acetic acid, lyophylized and subsequently used for immunization.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56
Test Specificity The antibody Ds-2 reacts with canine intra-acrosomal sperm protein (92 and 42 kDa), a testis-specific acrosomal vesicle protein 1 associated with the acrosomal membranes and matrix of the mature sperm.
Target Species Canine
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.