Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Intron-Binding Protein Of 160 KDa Antibody, Novus Biologicals™
SDP

Catalog No. NB035253 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB035253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB035253 Supplier Novus Biologicals Supplier No. NBP287631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Intron-Binding Protein Of 160 KDa Polyclonal specifically detects Intron-Binding Protein Of 160 KDa in Human samples. It is validated for Western Blot.

Specifications

Antigen Intron-Binding Protein Of 160 KDa
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias AQR, aquarius homolog (mouse), FSAP164, Functional Spliceosome-Associated Protein 164, IBP160, Intron-Binding Protein Aquarius
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of Human Intron-Binding Protein Of 160 KDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9716
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.