Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Intron-Binding Protein Of 160 KDa Antibody, Novus Biologicals™
SDP

Catalog No. NB019354 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019354 0.025 mg
NBP22360705 0.05 mg
NBP223607 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB019354 Supplier Novus Biologicals Supplier No. NBP2236070.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Intron-Binding Protein Of 160 KDa Polyclonal specifically detects Intron-Binding Protein Of 160 KDa in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Intron-Binding Protein Of 160 KDa
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Gene Accession No. O60306
Gene Alias AQR, aquarius homolog (mouse), FSAP164, Functional Spliceosome-Associated Protein 164, IBP160, Intron-Binding Protein Aquarius
Gene Symbols AQR
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the following peptide sequence: DYILLSLVRTRAVGHLC
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9716
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.