Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INTS1 Antibody, Novus Biologicals™
SDP

Catalog No. NB100509 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100509 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100509 Supplier Novus Biologicals Supplier No. NB100509
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

INTS1 Polyclonal specifically detects INTS1 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen INTS1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:2500, Immunoprecipitation 2 - 5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. XP_291222.2
Gene Alias DKFZp586J0619, FLJ46624, integrator complex subunit 1, NET28, UNQ1821/PRO3434
Gene Symbols INTS1
Host Species Rabbit
Immunogen A synthetic peptide derived from a sequence between residues 150 and 200 of human KIAA1440 using the numbering given in entry XP_291222.2 (GeneID 26173).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26173
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.