Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INTS8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100578
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5100578 100 μL
1 options

Catalog No. PIPA5100578

Supplier: Invitrogen™ PA5100578

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total INTS8.

This gene encodes a component of prolyl 4-hydroxylase, a key enzyme in collagen synthesis composed of two identical alpha subunits and two beta subunits. The encoded protein is one of several different types of alpha subunits and provides the major part of the catalytic site of the active enzyme. In collagen and related proteins, prolyl 4-hydroxylase catalyzes the formation of 4-hydroxyproline that is essential to the proper three-dimensional folding of newly synthesized procollagen chains. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INTS8
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene INTS8
Gene Accession No. Q75QN2
Gene Alias C8orf52; FLJ20530; Int8; integrator complex subunit 8; INTS8; MGC131633; Protein kaonashi-1
Gene Symbols INTS8
Host Species Rabbit
Immunogen A synthesized peptide derived from human INTS8(Accession Q75QN2), corresponding to amino acid residues C893-Y943.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55656
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.