Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Platinum™ SuperFi™ PCR Master Mix

Catalog No. p-6698556
Encompass_Preferred
Click to view available options
No. of Reactions:
100 Reactions
5 x 500 Reactions
500 Reactions

Invitrogen Platinum SuperFi PCR Master Mix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification.

Invitrogen Platinum SuperFi PCR Master Mix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification. Only template, primers, and water need to be added, thus reducing the number of pipetting steps during PCR reaction setup.

Platinum SuperFi PCR Master Mix retains all the features of Platinum SuperFi DNA Polymerase, a proofreading DNA polymerase that combines superior fidelity with trusted Platinum hot-start technology. Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis, and other applications benefiting from supreme sequence accuracy.

Platinum SuperFi DNA Polymerase is engineered with a DNA-binding domain resulting in high processivity and increased resistance to PCR inhibitors. This feature also enables fast-cycling protocols and amplification of long targets. The Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing non-specific amplification and primer degradation.

Features

  • Exceptional >300X Taq fidelity
  • High specificity and increased yields with Platinum hot-start technology
  • Robust amplification of difficult targets of high GC content
  • Room temperature reaction setup and 24-hour bench top stability of pre-assembled reactions
  • Amplification of samples with suboptimal purity

Applications

  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates
  • Template generation for sequencing
  • High-throughput PCR
  • Long PCR

Notes

  • Annealing rules for Platinum SuperFi DNA Polymerase are different from many common DNA polymerases.
  • Platinum SuperFi PCR Master Mix is supplied with separate SuperFi GC Enhancer for difficult and GC-rich templates (˃65% GC).
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 2X Platinum SuperFi PCR Master Mix (2 x 1.25 mL)
• SuperFi GC Enhancer (1.25 mL)
• Nuclease-free water (2 x 1.25 mL)

Store at –5°C to –30°C.
GC-Rich PCR Performance High
Polymerase Platinum SuperFi DNA Polymerase
Reaction Speed Fast
Quantity 100 reactions
Shipping Condition Wet or Dry Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq) 300X
Hot Start Built-In Hot Start
No. of Reactions 100 Reactions
Overhang Blunt
Reaction Format SuperMix or Master Mix
Size (Final Product) 20 kb or less
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.