Learn More
Invitrogen™ AccuPrime™ GC-Rich DNA Polymerase
AccuPrime GC-Rich DNA Polymerase is designed to provide high-yield, high-specificity amplification of difficult-to-amplify templates, such as those with >65% GC content.
$650.00 - $2671.65
Specifications
GC-Rich PCR Performance | High |
---|---|
Polymerase | AccuPrime GC-Rich DNA Polymerase |
Reaction Speed | Standard |
Shipping Condition | Wet or Dry Ice |
For Use With (Application) | Hot-start PCR |
Includes: AccuPrime GC-Rich DNA Polymerase (100μL, at 2U/μL) is supplied with two 5X reaction buffers and a separate vial (1mL) of 50mM MgSO4 . Both buffers contain dNTPs, MgSO4 , and AccuPrime accessory proteins. Reaction buffer A (1mL) is for genomic targets. Reaction buffer B (1mL) is for use with cDNA, plasmid, or lambda targets. Sufficient reagents are provided for 200 amplification reactions of 25μL each (at 1 unit of enzyme per reaction).
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
12-337-016
![]() |
Invitrogen™
12337016 |
200 Reactions |
Each for $650.00
|
|
|||||
12-337-024
![]() |
Invitrogen™
12337024 |
1000 Reactions |
Each
for $2,671.65
Save 7% Reg : |
![]() |
|||||
Description
AccuPrime GC-Rich DNA Polymerase is designed to provide high-yield, high-specificity amplification of difficult-to-amplify templates, such as those with >65% GC content. The kit offers a choice of buffers for amplifying genomic DNA targets (Buffer A) or non-GC-rich cDNA, plasmid, and lambda-based targets (Buffer B).
AccuPrime GC-Rich DNA Polymerase features
- High yields for targets up to 5 kb in length
- AccuPrime accessory proteins for improved PCR specificity
- Sensitivity down to 5 ng of template DNA
Robust and specific amplifications
The AccuPrime buffers contain thermostable proteins that enhance primer-template hybridization during PCR, increasing the specificity of the reaction. The polymerase itself, from the archaebacterium Pyrolobus fumarius, has a five-fold better processivity than Taq DNA polymerase, and remains active even after 4 hours at 95°C.
Unit definition
One unit of AccuPrime GC-Rich DNA Polymerase is the amount of enzyme required to incorporate 10 nmoles of dNTPs into acid-insoluble material in 30 min. at 74°C.
Order Info
Approved for shipment on Wet or Dry Ice
Specifications
High | |
Standard | |
Hot-start PCR | |
Built-In Hot Start | |
Separate Components |
AccuPrime GC-Rich DNA Polymerase | |
Wet or Dry Ice | |
2X | |
Blunt | |
5 kb or less |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.