Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNA Polymerase I, Large (Klenow) Fragment

Catalog No. 18012039
Encompass_Preferred
Click to view available options
Quantity:
100 Units
500 Units

Catalog No. 18-012-039

Supplier: Invitrogen™ 18012039

Only null left
Add to Cart
Add to Cart

Includes

DNA Polymerase I, Large (Klenow) Fragment is supplied with a vial of 10X REact™ 2 Buffer [500mM Tris-HCl (pH 8.0), 100mM MgCl2, 500mM NaCl], vial of dilution buffer.

DNA polymerase enzyme that lacks the 5' to 3' exonuclease activity of intact DNA Polymerase I, but does exhibit the 5' to 3' DNA polymerase and 3' to 5' exonuclease activities

Applications:
Fill-in of 5' overhangs . Synthesis of probes by random primers labeling method. Sequencing single- and double-stranded DNA. Site-directed mutagenesis.

Source:
Purified from E. coli expressing the Klenow fragment on a plasmid.

Performance and Quality Testing:
SDS-PAGE purity; single-stranded and double-stranded endodeoxyribonuclease and self-priming assays; performance evaluated in fill-in reaction.

Unit Definition:
One unit incorporates 10nMol of total deoxyribonucleotide into acid-precipitable material in 30 min. at 37°C using a template•primer.

Array CGH, Cloning, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Restriction Enzyme Cloning

Order Info

Shipping Condition: Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage DNA Polymerase I, Large (Klenow) Fragment is supplied with a vial of 10X REact™ 2 Buffer [500 mM Tris-HCl (pH 8.0), 100 mM
MgCl2 , 500 mM NaCl], vial of dilution buffer. Store at -20°C.
Description DNA Polymerase
Polymerase DNA Polymerase I
Exonuclease Activity 3' to 5'
Quantity 500 Units
Shipping Condition Approved for shipment on Wet or Dry Ice
Hot Start No
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.