Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Platinum™ Quantitative PCR SuperMix-UDG

Is a ready-to-use mixture for high-specificity, real-time quantitative DNA amplification, containing UDG to prevent carryover contamination (1,2).

$490.00 - $2165.00

Specifications

Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method qPCR
Reaction Speed Standard
For Use With (Equipment) 7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ
View More Specs

Includes: 2X concentration [40mM Tris-HCl (pH 8.4), 100mM KCl, 6mM MgCl2 , 400μM dATP, 400μM dCTP, 400μM dGTP, 800μM dUTP, 40 units/mL UDG, 60 units/ml Platinum™ Taq DNA Polymerase, stabilizers], vial of 50mM MgCl2

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
11-730-017
Encompass_Preferred
View Documents
Invitrogen™
11730017
100 Reactions
Each for $490.00
Only null left
Edge
Add to Cart
 
11-730-025
Encompass_Preferred
View Documents
Invitrogen™
11730025
500 Reactions
Each for $2,165.00
Only null left
Add to Cart
 
Description

Description

  • Highly sensitive and specific
  • Allows room temperature reaction assembly
  • Easy to use: just add template and fluorogenic primers or probes for detection
  • Includes a vial of 50mM MgCl2 to accommodate a variety of primer/template PCR systems
  • Compatible with most real-time instruments

Order Info

Guaranteed stable for 6 months when properly stored. Guaranteed minimum shelf life is 60 days. Shipping Conditions: Dry Ice.

Specifications

Specifications

Primer-probe
qPCR
7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ
Quantitative PCR SuperMix-UDG
Gene Expression
High
Standard
Platinum™
Dry Ice
DNA (Genomic), cDNA
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.