Learn More
Invitrogen™ SuperScript™ III Platinum™ One-Step qRT-PCR Kit w/ROX
One-Step qRT-PCR Kits with ROX combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression.
Supplier: Invitrogen™ 11745500

Includes: SuperScript™ III/Platinum™ Taq Mix, 2X Reaction Mix with ROX and 1mL Magnesium Sulfate (50mM). Guaranteed stable for 6 months when properly stored.
Description
One-Step qRT-PCR Kits with ROX combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient, high-throughput one-step format. One-Step qRT-PCR kits with ROX include SuperScript™ III Reverse Transcriptase which produces higher yields of full-length cDNA for broader gene representation, Platinum™ Taq DNA Polymerase which activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification and ROX reference dye premixed at optimal concentration for ABI instruments
Two kit configurations allow your choice of detection methods:- SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit provides a highly efficient, cost-effective qRT-PCR system with SYBR Green I detection
- SuperScript™ III Platinum™ One-Step qRT-PCR Kit provides sensitive, specific detection for use with LUX™ fluorogenic primers or dual-labeled probes (e.g. TaqMan™)
Order Info
Dry Ice

Specifications
• 500 μL SuperScript™ III/Platinum™ Taq Mix • 12.5 mL 2X Reaction Mix with ROX • 2 × 1 mL Magnesium Sulfate (50 mM) Store components in the dark at -20°C. |
|
1-step RT-qPCR | |
Standard | |
Platinum™, SuperScript™ | |
500 reactions | |
500 Reactions | |
500 Reactions |
Primer-probe | |
Platinum Taq DNA Polymerase | |
7000 System, 7300 System, 7700 System, 7900HT System | |
One-Step qRT-PCR Kit | |
Dry Ice | |
Gene Expression, Pathogen Detection, Virus Detection | |
RNA |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.