Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript™ Double-Stranded cDNA Synthesis Kit

Catalog No. 11917010
Encompass_Preferred
Click to view available options
Quantity:
10 Reactions
50 Reactions

Includes

Reagents and buffers for 10 first- and second-strand cDNA synthesis reactions converting 25-50μg total RNA or 0.2-5μg mRNA into double-stranded cDNA.

Requires

Primer

Maximize the yield of full-length cDNA, as well as that of overall cDNA synthesis

Contains all of the reagents (except an oligo (dT)-containing primer) necessary to make high-quality, double-stranded cDNA from total RNA or poly A+-selected RNA (mRNA).

Simple, reliable, and trouble-free
The SuperScript Double-Stranded cDNA Synthesis Kit offers a simple, reliable, and trouble-free method by which RNA templates can be converted to cDNA for use in cloning and library construction experiments. The reactions described in the SuperScript Double-Stranded cDNA Synthesis Kit protocol are designed to convert 25–50μg total RNA, or 0.2–5μg mRNA, into first- and second-strand cDNA (supply your own primer). Enjoy the benefits of a tested protocol and reagents of consistent quality. To get the best possible cDNA product using this kit, use a high-quality RNA substrate.

  • Reliability: each buffer, reagent, and enzyme in the kit is of the same high quality you expect from Life Technologies
  • Performance: SuperScript II is an enzyme you can trust for high-yield, high-quality, general-purpose, first-strand synthesis
  • Optimal temperature: 42°C

DNA and RNA Purification and Analysis, DNA Labeling, Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, Nucleic Acid Labeling and Oligo Synthesis, PCR and Real-Time PCR, Reverse Transcription

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contents: The kit includes reagents and buffers for 10 first- and second-strand cDNA synthesis reactions converting 25-50 μg total RNA or 0.2-5 μg mRNA into double-stranded cDNA.

Storage: Store at -20°C. All components are guaranteed stable for 6 months when properly stored.
Format Kit
Labeling Method Direct Labeling
Optimal Reaction Temperature 42°C
Product Line SuperScript™
Product Type cDNA Synthesis Kit
Quantity 10 Reactions
Reverse Transcriptase SuperScript™ II
Shipping Condition Wet Ice
For Use With (Application) Microarray Analysis
Final Product Type cDNA (Double-Stranded)
Sample Type RNA
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.