Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ T4 DNA Ligase (1 U/μL)

Catalog No. p-4918234
Encompass_Preferred
Click to view available options
Quantity:
100 U
4 x 500 U
500 U

Includes

Vial of 5X reaction buffer (250mM Tris-HCl (pH 7.6), 50mM MgCl2, 5mM ATP, 5mM DTT, 25% (w/v) polyethylene glycol-8000)

Ideal for cloning (blunt-end or cohesive-end ligation) and adding linkers or adapters to blunt-ended DNA

T4 DNA Ligase catalyzes the formation of phosphodiester bonds in the presence of ATP between double-stranded DNAs with 3'hydroxyl and 5'phosphate termini. Single-stranded nucleic acids are not substrates for this enzyme.

  • Optimizes ligation in 5 minutes
  • Source: Purified from E. coli lambda lysogen NM989
  • Performance and quality testing: Endodeoxyribonuclease, 3' and 5' exodeoxyribonuclease assays; ligation efficiency tested
  • Unit definition: One unit catalyzes exchange of 1nmol 32P-labeled pyrophosphate into ATP in 20 min. at 37°C. One unit is equal to approximately 300 cohesive-end ligation units
  • Unit reaction conditions: 66mM Tris-HCl (pH 7.6), 6.6mM MgCl2, 10mM DTT, 66μM ATP, 3.3μM 32P-labeled pyrophosphate, and enzyme in 0.1mL for 20 min. at 37°C

ChIP-on-Chip, Chromatin Biology, Cloning, RNAi, Epigenetics & Noncoding RNA Research, Restriction Enzyme Cloning

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 U/μL
Content And Storage T4 DNA Ligase is supplied with a vial of 5X reaction buffer [250 mM Tris-HCl (pH 7.6), 50 mM MgCl2, 5 mM ATP, 5 mM DTT, 25% (w/v) polyethylene glycol-8000]. Store at -20°C.
Shipping Condition Dry Ice
Enzyme Ligase
Compatible Buffer DNA Ligase Buffer, 5X Reaction Buffer
Quantity 100 U
Product Type T4 DNA Ligase
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.