Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Involucrin Antibody (IVRN/2113R), HRP, Novus Biologicals™
SDP

Catalog No. NB392368 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392368 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392368 Supplier Novus Biologicals Supplier No. NBP308386H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Involucrin Monoclonal antibody specifically detects Involucrin in Human, Porcine, Canine, Primate, Monkey, Mouse (Negative) samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Involucrin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IVRN/2113R
Conjugate HRP
Formulation PBS
Gene Alias involucrin
Host Species Rabbit
Immunogen Recombinant human full-length Involucrin protein (IVRN) (Uniprot: P07476)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 3713
Target Species Human, Pig, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.