Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Involucrin Antibody (SY8), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N102774A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N102774A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N102774A750 Supplier Novus Biologicals Supplier No. NB1002774AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Involucrin Monoclonal specifically detects Involucrin in Canine, Human, Pig, Primate samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen Involucrin
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone SY8
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias involucrin
Gene Symbols IVL
Host Species Mouse
Immunogen Purified involucrin from human keratinocytes.
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3713
Target Species Canine, Human, Pig, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.