Iodine, resublimed crystals, 99.9985% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1061988

Quantity 2.5g

 View more versions of this product

Catalog No. AA1061988

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7553-56-2
    Physical Form Resublimed Crystals
    Assay Percent Range 99.9985% (Metals basis)
    Molecular Formula I2
    Formula Weight 253.81
    MDL Number MFCD00011355
    Chemical Name or Material Iodine

    Specifications

    Physical Form Resublimed Crystals
    Formula Weight 253.81
    Quantity 2.5g
    Color Blue-black
    DOT Information DOT Class: 8, Packaging Group: III
    EINECS Number 231-442-4
    Odor Irritating
    Assay Percent Range 99.9985% (Metals basis)
    Molecular Formula I2
    Appearance Bluish-black crystals
    MDL Number MFCD00011355
    CAS 7553-56-2
    Health Hazard 1 H400-H312-H332: Very toxic to aquatic life. Harmful in contact with skin. Harmful if inhaled.
    Chemical Name or Material Iodine

    H400-H312-H332: Very toxic to aquatic life. Harmful in contact with skin. Harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit