Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine Solution, 0.1N (N/10) (Certified), Fisher Chemical™

Catalog No. p~213447 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SI86-1 1 L Glass Bottle
SI864 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SI86-1

Standardized volumetrically at 25°C against NIST™ arsenous acid

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0 , 7553-56-2 , 7732-18-5
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011355
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Color Amber
Boiling Point 100°C
Melting Point 0°C
Concentration 0.0995 to 0.1005N
Density 1.0g/cm³
Grade Certified
Concentration or Composition (by Analyte or Components) 0.1N (N/10)
Identification Pass test
Packaging Glass Bottle
Traceability to NIST Lot number used arsenic trioxide
Vapor Pressure 14mmHg
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Iodine Solution, 0.1N (N/10)
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.