Learn More
Iodoacetic acid, 97+%
Description
Iodoacetic acid is used as a reagent for the modification of sulfhydryl groups in organic synthesis. It reacts with cysteine moiety in proteins to prevent the re-formation of disulfide bonds during protein sequencing. It is also used as a general reagent in acylation reactions as well as in glycopeptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Iodoacetic acid is used as a reagent for the modification of sulfhydryl groups in organic synthesis. It reacts with cysteine moiety in proteins to prevent the re-formation of disulfide bonds during protein sequencing. It is also used as a general reagent in acylation reactions as well as in glycopeptide synthesis.
Solubility
Soluble in water, alcohol and ethanol. Slightly soluble in ether and chloroform.
Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents, bases and strong reducing agents.
Specifications
Specifications
| Melting Point | 79°C to 82°C |
| Boiling Point | 208°C (decomposition) |
| Odor | Pungent |
| Quantity | 500 g |
| UN Number | UN2923 |
| Beilstein | 1739079 |
| Merck Index | 14,5025 |
| Formula Weight | 185.95 |
| Percent Purity | ≥97% |
| Chemical Name or Material | Iodoacetic acid |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H301-H314 Toxic if swallowed. Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S. |
| RTECSNumber | AI3500000 |
| TSCA | TSCA |
| Recommended Storage | 2°C to 8°C |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.