Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iododeoxyuridine/IdU Antibody (32D8.D9), Novus Biologicals™
SDP

Catalog No. NBP244056 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244056 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP244056 Supplier Novus Biologicals Supplier No. NBP244056
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Ensure accurate, reproducible results in Western Blot, Dot Blot, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry

Iododeoxyuridine/IdU Monoclonal specifically detects Iododeoxyuridine/IdU in Human, Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Iododeoxyuridine/IdU
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Dot Blot
Classification Monoclonal
Clone 32D8.D9
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:5000, Flow Cytometry 1:10-1:1000, ELISA 1:1000-1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Alias 2'-Deoxy-5-iodouridine, 54-42-2, 5-Iodo-2'-deoxyuridine, 5-Iododeoxyuridine, Idoxuridin, Iododeoxyridine, Iodoxuridine, Joddeoxiuridin
Host Species Mouse
Immunogen Iododeoxyuridine/Iododeoxyuridine/IdU Antibody (32D8.D9) was produced in mice by repeated immunizations prepared via immunizations with Bromodeoxyuridine-KLH followed by hybridoma development.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific for Iododeoxyuridine/IdU. Cross-reactivity is not observed with BrdU, CldU or FdU.
Target Species Non-species specific
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.