Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IP6K1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB329285 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329285 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329285 Supplier Novus Biologicals Supplier No. NBP300228PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IP6K1 Polyclonal antibody specifically detects IP6K1 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen IP6K1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate PE
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias ATP:1D-myo-inositol-hexakisphosphate phosphotransferase, EC 2.7.4.21, inositol hexakisphosphate kinase 1, Inositol hexaphosphate kinase 1KIAA0263IHPK1PiUS, InsP6 kinase 1, MGC9925, Pi uptake stimulator
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human IP6K1 (Uniprot#: Q92551-1; Met1-Gln441)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9807
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.