Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IPCEF1 Antibody (OTI3G2), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP271781A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271781A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271781A70 Supplier Novus Biologicals Supplier No. NBP271781AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IPCEF1 Monoclonal specifically detects IPCEF1 in Human samples. It is validated for Western Blot.

Specifications

Antigen IPCEF1
Applications Western Blot
Classification Monoclonal
Clone OTI3G2
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias interaction protein for cytohesin exchange factors 1, interactor protein for cytohesin exchange factors 1, KIAA0403interaction protein for cytohesin exchange factors 1 Interaction protein forcytohesin exchange factors 1, phosphoinositide binding protein PIP3-E, Phosphoinositide-binding protein PIP3-E, PIP3-E, RP3-402L9.2
Gene Symbols IPCEF1
Host Species Mouse
Immunogen Full length recombinant protein of human IPCEF1 (NP_056368) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 26034.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.