Learn More
Description
- Eliminates background staining
- Use with Immobiline DryStrip gels to improve protein solubility
- Ensures optimal conditions for IEF separation
- High sample loading capacity, improved resolution, and increases the probability of detecting low abundance proteins when used with Immobiline DryStrip gels
Order Info
Supplied in 1mL aliquots of water; order Immobiline Drystrip gels separately.
Specifications
Specifications
| Color | Colorless to Light Yellow |
| pH Range | 4 to 7 |
| Form | Liquid |
| For Use With (Application) | IEF, 2-D Electrophoresis |
| Quantity | 1mL |
| Storage Requirements | 2° to 8°C |
| Product Type | IPG Buffer |
| For Use With (Equipment) | Immobiline DryStrip Gels |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.