Learn More
Description
- Eliminates background staining
- Use with Immobiline DryStrip gels to improve protein solubility
- Ensures optimal conditions for IEF separation
- High sample loading capacity, improved resolution, and increases the probability of detecting low abundance proteins when used with Immobiline DryStrip gels
Order Info
Supplied in 1mL aliquots of water; order Immobiline Drystrip gels separately.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Color | Colorless to Light Yellow |
| pH Range | 3 to 10 nonlinear |
| Form | Liquid |
| For Use With (Application) | IEF, 2-D Electrophoresis |
| Quantity | 1mL |
| Storage Requirements | 2° to 8°C |
| Product Type | IPG Buffer |
| For Use With (Equipment) | Immobiline DryStrip Gels |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
