Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ipilimumab Mouse, Clone: 26B6H7D9, GenScript™

Catalog No. 502120625
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μg
Catalog No. Quantity
50-212-0625 40 μg
1 options

Catalog No. 50-212-0625

Supplier: Genscript Corporation A0185940

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/mL. Ipilimumab (Yervoy) is a humanized monoclonal antibody that is approved by the U.S. Food and Drug Administration for the treatment of patients with unresectable or metastatic melanoma. Ipilimumab works to activate the immune system by targeting CTLA-4 (Cytotoxic T-lymphocyte protein 4), a negative regulator of cytotoxic T-lymphocyte activity. Ipilimumab binds to CTLA-4, blocking the inhibitory signal, which allows the cytotoxic T-lymphocytes to destroy the cancer cells Anti-Ipilimumab Antibody (26B6H7D9), mAb, Mouse is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a mouse immunized with Ipilimumab.

Ipilimumab Monoclonal antibody specifically detects Ipilimumab in Chemical samples. It is validated for ELISA

Specifications

Antigen Ipilimumab
Applications ELISA
Classification Monoclonal
Clone 26B6H7D9
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene Alias Yervoy
Host Species Mouse
Immunogen Ipilimumab
Purification Method Protein A
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.