Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IPO-38 Antibody (SPM515) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB121580 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB121580 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB121580 Supplier Novus Biologicals Supplier No. NBP311493
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IPO-38 Monoclonal antibody specifically detects IPO-38 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence

Specifications

Antigen IPO-38
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone SPM515
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Formulation 10 mM PBS
Host Species Mouse
Immunogen Spleen cells of a patient with hairy cell leukemia
Purification Method Bioreactor concentrate
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.