Learn More
Honeywell Analytics™ IQ Express Docking Stations
Automatic calibration station and data download center compatible with Sperian gas detectors
Supplier: Honeywell Analytics™ 54469100
Includes: Software, installation disks, and manuals.
Requires : Appropriate calibration gas for bump tests and span calibrations.
Description
Just drop the detector into the dock, and the IQ Express does the rest—in as little as 20 seconds.
- Identifies instrument and performs a battery check before moving on to bump test
- Performs full span calibration if bump test fails; no user intervention is required
- Tests audible, visible, and vibrating alarms
- Downloads instrument data
- Displays the word “Ready” when all tests and functions are completed
Order Info
Standard stations are suitable for detectors outfitted with O2, LEL, CO, or H2S sensors. Enhanced versions are suitable for those sensors, as well as SO2, Cl2, NO2, NH3, HCN and PH3.
Specifications
Oxygen, Carbon Monoxide, Hydrogen Sulfide, Sulfur Dioxide, Chlorine, Nitrogen Dioxide, Ammonia, Hydrogen Cyanide, Phosphine | |
Toxi Pro and ToxiLtd Personal Gas Detectors | |
Docking Station |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.