Learn More
Honeywell Analytics™ IQ Express Docking Stations
Automatic calibration station and data download center compatible with Sperian gas detectors
$7252.20
Specifications
Type Docking Station
View More Specs
Requires: Appropriate calibration gas for bump tests and span calibrations.
Products 1
Catalog Number Mfr. No. For Use With (Equipment) Components Measured Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Components Measured Price Quantity & Availability  
19-150-1201
Encompass
View Documents
Honeywell Analytics™
54469100N
ToxiPro/LTD Oxygen, Carbon Monoxide, Hydrogen, Sulfur, Sulfur Dioxide, Chlorine, Nitrogen Dioxide
Each of 1 for $7,252.20
Only null left
Add to Cart
Description
Description
Just drop the detector into the dock, and the IQ Express does the rest—in as little as 20 seconds.
  • Identifies instrument and performs a battery check before moving on to bump test
  • Performs full span calibration if bump test fails; no user intervention is required
  • Tests audible, visible, and vibrating alarms
  • Downloads instrument data
  • Displays the word “Ready” when all tests and functions are completed
Order Info
Standard stations are suitable for detectors outfitted with O2, LEL, CO, or H2S sensors. Enhanced versions are suitable for those sensors, as well as SO2, Cl2, NO2, NH3, HCN and PH3.
Specifications
Specifications
Docking Station
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.