Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ iQ Series™ Men's Comfort Knit Red Flame Resistant Henley

Catalog No. 17103753 Shop All Red Kap Industries Products
Click to view available options
Size:
Large (Long)

iQ Series™ Men's Comfort Knit Flame Resistant Henley is CAT 2 AR/FR, NFPA™ 2112 compliant and a casual, comfortable and stylish part of your FR system.

  • With raglan sleeves and underarm gussets for added mobility, so you can bend, reach, stretch and work better
  • One concealed chest pocket with pencil stall and one concealed pencil stall sleeve pocket
  • Hidden rib knit cuffs to keep sleeves in place
  • Moisture-wicking, breathable fabric
  • Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Flash Fire, Arc Flash
  • Wash Care: Home Wash
  • Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame resistant, 5.3 oz. (180 g/m2)
  • Blend: 69% Cotton/25% Polyester/6% Polyoxadiazole
  • Closure: Four-button placket
  • Product Collection: iQ Series Comfort, iQ Series
  • Packaging: Polybag

Specifications

Size Large (Long)
Gender Men's
Material Cotton, Polyester, POD
Color Red
Neck Style Henley
Sleeve Style Raglan Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 2
Hazard Risk Category Category 2
Certifications/Compliance ASTM F1506, NFPA 2112, NFPA 70E
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.