Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IQGAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589674
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA589674 100 μL
1 options

Catalog No. PIPA589674

Supplier: Invitrogen™ PA589674

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QRRVLRNLRR LEALGLVSAR NGYQGLVDEL AKDIRNQHRH RHRRKAELVK LQATLQGLST KTTFYEEQGD YYSQYIRACL DHLAPDSKSS GKGKKQPSLH YTAAQLLEKG VLVEIEDLPA SHFRNVIFDI TPGDEAGKFE VNAKFLGVDM ERFQLHYQDL LQLQYEGVAV MKLFNKAKVN VNLLIFLLNK KFLRK; Positive Samples: 293T.

IQGAP3 (IQ motif containing GTPase activating protein 3) is a 1,631 amino acid protein that acts as an effector of Cdc42 and Rac 1, linking their activation to the cytoskeleton during neuronal morphogenesis. A novel member of the IQGAP family, IQGAP3 is highly expressed in brain where it localizes to axons of hippocampal neurons. IQGAP3 contains one Ras-GAP domain, a CH (calponin-homology) domain, four IQ domains and is encoded by a gene located on human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome. Chromosome 1 houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson's disease, Gaucher disease, schizophrenia and Usher syndrome. Aberrations in chromosome 1 are found in a variety of cancers, including head and neck cancer, malignant melanoma and multiple myeloma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IQGAP3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IQGAP3
Gene Accession No. Q86VI3
Gene Alias AI593484; D030034H08; IQ motif containing GTPase activating protein 3; IQGA3; IQGAP3; MGC10170; Ras GTPase-activating-like protein IQGAP3
Gene Symbols IQGAP3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1437-1631 of human IQGAP3 (NP_8399432).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128239, 310621, 404710
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.