Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IQSEC1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB341714 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341714 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341714 Supplier Novus Biologicals Supplier No. NBP281958MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IQSEC1 Polyclonal antibody specifically detects IQSEC1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen IQSEC1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ADP-ribosylation factors guanine nucleotide-exchange protein 100, ADP-ribosylation factors guanine nucleotide-exchange protein 2, ARFGEP100, BRAG2, IQ motif and Sec7 domain 1, IQ motif and SEC7 domain-containing protein 1, KIAA0763Brefeldin-resistant Arf-GEF 2 protein
Host Species Rabbit
Immunogen IQSEC1 antibody was raised against a 19 amino acid peptide near the amino terminus of human IQSEC1. The immunogen is located within amino acids 60 - 110 of IQSEC1. Amino Acid Squence: PPGQQQRTRRPKLQHSTSI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9922
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.